Search
Search

Habp2| Gene Symbol: Habp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146101.1 | NP_666213.1 | 6 - 7 | 667 | 54 | |
| GenBank mRNA | AK128915.1 | - | 7 - 8 | 793 | 54 | |
| AK132407.1 | - | 684 | 54 | |||
| BC031775.1 | - | 6 - 7 | 667 | 4242577 | 54 | |
Set Membership: |
|