Search
Search
| Gene Symbol: Alg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081264.1 | NP_001074733.1 | 5 - 6 | 545 | 65 | |
| GenBank mRNA | AK157903.1 | - | 5 - 6 | 507 | 65 | |
| AK171887.1 | - | 5 - 6 | 498 | 65 | ||
| BC050854.1 | - | 5 - 6 | 510 | 6332345 | 65 | |
| BC080867.1 | - | 5 - 6 | 485 | 65 | ||
Set Membership: |
|