Search
Search

Nop58| Gene Symbol: Nop58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018868.2 | NP_061356.2 | 8 - 9 | 955 | 135 | |
| GenBank mRNA | AF053232.1 | - | 8 - 9 | 860 | 135 | |
| AK044216.1 | - | 952 | 135 | |||
| AK133959.1 | - | 8 - 9 | 953 | 135 | ||
| AK134286.1 | - | 8 - 9 | 956 | 135 | ||
| AK212676.1 | - | 2 - 3 | 254 | 135 | ||
| BC076604.1 | - | 8 - 9 | 956 | 30630088 | 135 | |
| BC085135.1 | - | 8 - 9 | 925 | 5718488 | 135 | |
Set Membership: |
|