Search
Search
| Gene Symbol: Ipo8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081113.1 | NP_001074582.1 | 17 - 18 | 2161 | 93 | |
| GenBank mRNA | BC034091.1 | - | 4 - 5 | 453 | 93 | |
| BC151036.1 | - | 17 - 18 | 2029 | 9087949 | 93 | |
| BC151052.1 | - | 17 - 18 | 2029 | 9087965 | 93 | |
Set Membership: |
|