Search
Search
| Gene Symbol: Aox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008419.2 | NP_001008419.1 | 23 - 24 | 2675 | 103 | |
| GenBank mRNA | AY187018.2 | - | 23 - 24 | 2675 | 103 | |
| AY665589.1 | - | 23 - 24 | 2582 | 103 | ||
| BC150826.1 | - | 23 - 24 | 2602 | 9087737 | 103 | |
Set Membership: |
|