Search
Search
| Gene Symbol: Aox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008419.2 | NP_001008419.1 | 24 - 25 | 2764 | 90 | |
| GenBank mRNA | AY187018.2 | - | 24 - 25 | 2764 | 90 | |
| AY665589.1 | - | 24 - 25 | 2671 | 90 | ||
| BC150826.1 | - | 24 - 25 | 2691 | 9087737 | 90 | |
Set Membership: |
|