Search
Search
| Gene Symbol: Miip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025365.2 | NP_001020536.2 | 7 - 8 | 1272 | 104 | |
| GenBank mRNA | AK133976.1 | - | 9 - 10 | 1424 | 104 | |
| AK170224.1 | - | 6 - 7 | 1394 | 104 | ||
| BC017525.1 | - | 6 - 7 | 1344 | 4910951 | 104 | |
| BC024069.1 | - | 7 - 8 | 1054 | 5097088 | 104 | |
| CO039431.1 | - | 3 - 4 | 104 | |||
| FJ618908.1 | - | 7 - 8 | 1272 | 104 | ||
Set Membership: |
|