Search
Search

Car14| Gene Symbol: Car14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011797.2 | NP_035927.1 | 5 - 6 | 760 | 116 | |
| GenBank mRNA | AB005450.1 | - | 5 - 6 | 760 | 116 | |
| AK012316.1 | - | 2 - 3 | 137 | 116 | ||
| AK157764.1 | - | 5 - 6 | 760 | 116 | ||
| BC046995.1 | - | 5 - 6 | 742 | 6491779 | 116 | |
Set Membership: |
|