Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021529.3 | NP_067504.2 | 4 - 5 | 517 | 117 | |
GenBank mRNA | AB041577.1 | - | 4 - 5 | 506 | 117 | |
AJ238247.2 | - | 3 - 4 | 344 | 117 | ||
AK013403.1 | - | 3 - 4 | 313 | 117 | ||
AK015746.1 | - | 3 - 4 | 346 | 117 | ||
AK028063.1 | - | 4 - 5 | 482 | 117 | ||
AK028254.1 | - | 3 - 4 | 436 | 117 | ||
AK033405.1 | - | 4 - 5 | 516 | 117 | ||
AK040589.1 | - | 3 - 4 | 345 | 117 | ||
AK087572.1 | - | 3 - 4 | 340 | 117 | ||
AK088240.1 | - | 3 - 4 | 426 | 117 | ||
AK163192.1 | - | 516 | 117 | |||
AK168298.1 | - | 4 - 5 | 519 | 117 | ||
BC024754.1 | - | 4 - 5 | 480 | 5375405 | 117 | |
BC138295.1 | - | 3 - 4 | 313 | 8861314 | 117 | |
BC138296.1 | - | 3 - 4 | 313 | 8861315 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |