Search
Search

Nmnat1| Gene Symbol: Nmnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133435.1 | NP_597679.1 | 1 - 2 | 34 | 63 | |
| GenBank mRNA | AK011137.1 | - | 1 - 2 | 56 | 63 | |
| AK017631.1 | - | 44 | 63 | |||
| AK152407.1 | - | 94 | 63 | |||
| AY679721.1 | - | 1 - 2 | 34 | 63 | ||
| BC138500.1 | - | 1 - 2 | 102 | 8861520 | 63 | |
| BC138502.1 | - | 1 - 2 | 102 | 8861522 | 63 | |
Set Membership: |
|