Search
Search
| Gene Symbol: Scp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011327.4 | NP_035457.1 | 4 - 5 | 436 | 70 | |
| GenBank mRNA | AK004860.2 | - | 4 - 5 | 366 | 70 | |
| BC018384.1 | - | 4 - 5 | 391 | 3710340 | 70 | |
| BC034613.1 | - | 4 - 5 | 357 | 5123611 | 70 | |
| M91458.1 | - | 4 - 5 | 351 | 70 | ||
Set Membership: |
|