Search
Search

Eps15| Gene Symbol: Eps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159964.1 | NP_001153436.1 | 2 - 3 | 465 | 139 | |
| NM_007943.3 | NP_031969.1 | 14 - 15 | 1414 | 139 | ||
| GenBank mRNA | AK083176.1 | - | 2 - 3 | 449 | 139 | |
| AY190610.1 | - | 14 - 15 | 1387 | 139 | ||
| BC048783.1 | - | 14 - 15 | 1388 | 6466448 | 139 | |
| CF533628.1 | - | 2 - 3 | 465 | 139 | ||
| L21768.1 | - | 14 - 15 | 1387 | 139 | ||
Set Membership: |
|