Search
Search
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 11 - 12 | 1499 | 78 | |
| GenBank mRNA | AK031548.1 | - | 11 - 12 | 1499 | 78 | |
| AK040377.1 | - | 10 - 11 | 1730 | 78 | ||
| AK166470.1 | - | 9 - 10 | 1498 | 78 | ||
| BC023786.1 | - | 10 - 11 | 877 | 78 | ||
| BC026832.1 | - | 11 - 12 | 1549 | 4486176 | 78 | |
| BC029977.1 | - | 2 - 3 | 135 | 78 | ||
| BC036128.1 | - | 11 - 12 | 1617 | 4971791 | 78 | |
Set Membership: |
|