Search
Search
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 19 - 20 | 2224 | 146 | |
| GenBank mRNA | AK031548.1 | - | 19 - 20 | 2224 | 146 | |
| AK166470.1 | - | 17 - 18 | 2223 | 146 | ||
| BC023786.1 | - | 18 - 19 | 1602 | 146 | ||
| BC024544.1 | - | 3 - 4 | 209 | 146 | ||
| BC026832.1 | - | 19 - 20 | 2274 | 4486176 | 146 | |
| BC029977.1 | - | 10 - 11 | 860 | 146 | ||
| BC036128.1 | - | 19 - 20 | 2342 | 4971791 | 146 | |
| U86112.1 | - | 5 - 6 | 472 | 146 | ||
Set Membership: |
|