Search
Search

Cdk11b| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007661.3 | NP_031687.2 | 12 - 13 | 1433 | 70 | |
| GenBank mRNA | AK077668.1 | - | 12 - 13 | 1434 | 70 | |
| AK147133.1 | - | 12 - 13 | 1411 | 70 | ||
| BC025058.1 | - | 1 - 2 | 83 | 70 | ||
| BC052920.1 | - | 12 - 13 | 1511 | 6489832 | 70 | |
| L37092.1 | - | 11 - 12 | 2005 | 70 | ||
| M58633.1 | - | 7 - 8 | 776 | 70 | ||
Set Membership: |
|