Search
Search

Casc1| Gene Symbol: Casc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177222.4 | NP_796196.3 | 7 - 8 | 739 | 65 | |
| GenBank mRNA | AK034148.1 | - | 6 - 7 | 587 | 65 | |
| AK132912.1 | - | 7 - 8 | 963 | 65 | ||
| AY423542.1 | - | 7 - 8 | 670 | 65 | ||
| AY485607.1 | - | 7 - 8 | 739 | 65 | ||
| AY485608.1 | - | 7 - 8 | 739 | 65 | ||
| BC072568.1 | - | 5 - 6 | 581 | 65 | ||
| BC120743.1 | - | 7 - 8 | 643 | 40129666 | 65 | |
| BC125423.1 | - | 7 - 8 | 643 | 40129938 | 65 | |
| BC144801.1 | - | 7 - 8 | 643 | 65 | ||
Set Membership: |
|