Search
Search

4921539E11Rik| Gene Symbol: 4921539E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163494.1 | NP_001156966.1 | 3 - 4 | 823 | 151 | |
| NM_027612.1 | NP_081888.1 | 4 - 5 | 915 | 151 | ||
| GenBank mRNA | AK015018.1 | - | 4 - 5 | 910 | 151 | |
| AK161421.1 | - | 4 - 5 | 915 | 151 | ||
Set Membership: |
|