| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P277071 |
BX631077.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P277071 |
NM_027612.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P277071 |
AK015018.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P277071 |
NM_001163494.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P277071 |
BX631077.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P277071 |
NM_027612.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P277071 |
AK015018.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P277071 |
NM_001163494.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969780 |
BC107404.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969780 |
BC107405.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969780 |
NM_027612.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969780 |
AK015018.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969780 |
AK161421.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514028 |
AK009576.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514028 |
AK032357.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514028 |
AK087101.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514028 |
AK155096.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514028 |
AK156895.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514028 |
AK172330.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514028 |
NM_001029990.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886719 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346186 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
150205 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
443635 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
237663 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
331480 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
235578 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376680 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391631 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776628 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
539418 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
114521 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
389514 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
627956 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
748184 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
433283 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
295270 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
800268 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798228 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
286043 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
537336 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479606 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
44032 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
885030 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
482717 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
10748 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
98643 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
795411 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
175656 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
668593 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
786023 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
5685 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
132868 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
65267 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2585 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
70283 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P103712 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P172694 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P277071 |
BX631077.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P277071 |
NM_027612.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P277071 |
AK015018.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P277071 |
NM_001163494.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P277071 |
BX631077.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P277071 |
NM_027612.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P277071 |
AK015018.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P277071 |
NM_001163494.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969780 |
BC107404.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969780 |
BC107405.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969780 |
NM_027612.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969780 |
AK015018.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969780 |
AK161421.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05340867 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05340877 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P103712 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P172694 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00946885 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05340932 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05340863 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00946858 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P277071 |
BX631077.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P277071 |
NM_027612.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P277071 |
AK015018.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P277071 |
NM_001163494.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P277071 |
BX631077.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P277071 |
NM_027612.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P277071 |
AK015018.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P277071 |
NM_001163494.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P103712 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05340932 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P172694 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05340877 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165342 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03899692 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165307 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165302 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165327 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03851448 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03334054 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03435464 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165145 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04062260 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03334052 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165308 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165322 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03435459 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165332 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165320 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165144 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03645572 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165067 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165339 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03808874 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165142 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165340 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03714824 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165325 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165326 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165299 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03621181 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165313 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165330 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165323 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165337 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165329 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165334 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23165147 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03808875 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03617771 |
- |
- |