Search
Search
| Gene Symbol: Tle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285529.1 | NP_001272458.1 | 5 - 6 | 1182 | 56 | |
| NM_001285530.1 | NP_001272459.1 | 5 - 6 | 1182 | 56 | ||
| NM_001285531.1 | NP_001272460.1 | 5 - 6 | 1182 | 56 | ||
| NM_001285532.1 | NP_001272461.1 | 5 - 6 | 1182 | 56 | ||
| NM_011599.5 | NP_035729.3 | 5 - 6 | 1182 | 56 | ||
| GenBank mRNA | AK046402.1 | - | 5 - 6 | 1150 | 56 | |
| AK048515.1 | - | 5 - 6 | 1168 | 56 | ||
| AK052961.1 | - | 3 - 4 | 1283 | 56 | ||
| AK076750.1 | - | 5 - 6 | 564 | 56 | ||
| AK082499.1 | - | 5 - 6 | 1148 | 56 | ||
| AY155195.1 | - | 5 - 6 | 341 | 56 | ||
| AY155196.1 | - | 5 - 6 | 1151 | 56 | ||
| AY155197.1 | - | 5 - 6 | 340 | 56 | ||
| AY155199.1 | - | 5 - 6 | 341 | 56 | ||
| AY155200.1 | - | 1 - 2 | 37 | 56 | ||
| BC057573.1 | - | 5 - 6 | 1012 | 6401650 | 56 | |
Set Membership: |
|