Search
Search
| Gene Symbol: Sae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285892.1 | NP_001272821.1 | 8 - 9 | 1101 | 63 | |
| GenBank mRNA | AK011772.1 | - | 8 - 9 | 1012 | 63 | |
| AK162789.1 | - | 8 - 9 | 989 | 63 | ||
Set Membership: |
|