Search
Search

Apex2| Gene Symbol: Apex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029943.1 | NP_084219.1 | 4 - 5 | 634 | 109 | |
| GenBank mRNA | AB072498.1 | - | 4 - 5 | 634 | 109 | |
| AK021248.1 | - | 4 - 5 | 109 | |||
| AK033489.1 | - | 4 - 5 | 626 | 109 | ||
| AK040145.1 | - | 4 - 5 | 568 | 109 | ||
| AK050858.1 | - | 4 - 5 | 597 | 109 | ||
| AK080916.1 | - | 846 | 109 | |||
| AK081677.1 | - | 4 - 5 | 589 | 109 | ||
| AK088918.1 | - | 4 - 5 | 603 | 109 | ||
| BC053107.1 | - | 3 - 4 | 1366 | 109 | ||
| BC078633.1 | - | 4 - 5 | 592 | 6488060 | 109 | |
Set Membership: |
|