Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01262729_m1
          See other PPP2R2B GE Assays ›
          Gene Symbol:
          Ppp2r2b
          Gene Name:
          protein phosphatase 2, regulatory subunit B, beta
          Gene Aliases:
          2900026H06Rik, 6330404L05Rik, E130009M08Rik, PP2A-PR55B, PR55-BETA, SCA12
          Chromosome Location:
          Chr.18: 42645221 - 43059471 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01262729_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp2r2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028392.3 NP_082668.1 3 - 4 572 71
          NR_073583.1 3 - 4 1024 71
          GenBank mRNA AF512670.1 - 3 - 4 278 71
          AF536771.1 - 3 - 4 341 71
          AK013600.1 - 3 - 4 574 71
          AK018119.1 - 3 - 4 529 71
          AK039592.1 - 3 - 4 415 71
          AK079059.1 - 1022 71
          AK148956.1 - 3 - 4 543 71
          BC026686.1 - 3 - 4 510 5065516 71
          BC066007.1 - 3 - 4 812 71
          BC085147.1 - 3 - 4 724 71
          BC088979.1 - 3 - 4 507 5690600 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001749.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp2r2b

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial fission
          protein targeting to mitochondrion
          apoptotic process
          signal transduction
          regulation of protein phosphatase type 2A activity
          positive regulation of neuron apoptotic process
          mitochondrial fragmentation involved in apoptotic process
          protein phosphatase type 2A regulator activity
          protein complex binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800009370.mm.1 NM_028392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009370.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009370.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009380.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009380.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009371.mm.1 NM_028392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009371.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009371.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009364.mm.1 NM_028392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009364.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009364.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009394.mm.1 NM_028392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009394.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009373.mm.1 NM_028392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009373.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009373.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009375.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009375.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009378.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009378.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009379.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009379.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009393.mm.1 NM_028392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009393.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009376.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009376.mm.1 TC1800001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009377.mm.1 NR_073583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009377.mm.1 TC1800001749.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 795200 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline