Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01263665_g1
          See other MFSD11 GE Assays ›
          Gene Symbol:
          Mfsd11
          Gene Name:
          major facilitator superfamily domain containing 11
          Gene Aliases:
          2600014M03Rik
          Chromosome Location:
          Chr.11: 116852283 - 116875660 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01263665_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfsd11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178620.3 NP_848735.1 12 - 13 1247 76
          GenBank mRNA AF015191.1 - 12 - 13 1213 76
          AK031384.1 - 12 - 13 1442 76
          AK032043.1 - 12 - 13 1443 76
          BC016209.1 - 13 - 14 1206 76
          BC079583.1 - 12 - 13 1274 76
          BC088988.1 - 12 - 13 1247 6418757 76
          BC100547.1 - 12 - 13 3323 76
          BC137970.1 - 12 - 13 1234 8860989 76
          BC137971.1 - 12 - 13 1235 8860990 76
          BC145068.1 - 12 - 13 1214 76

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mfsd11

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC137970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC137971.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC145068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC079583.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC088988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC137970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC137971.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC145068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC079583.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238734 BC088988.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760172 AF030131.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760172 AK045470.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760172 AK173185.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760172 BC060113.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760172 BC060696.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760172 NM_021506.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 515097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135237 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128150 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC137970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC137971.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC145068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC079583.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC088988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC137970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC137971.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC145068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC079583.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238734 BC088988.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28161659 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128150 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07506571 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07506586 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07506637 - -
          Whole Mouse Genome G4122A A_51_P238734 BC137970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC137971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC145068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC079583.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC088988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC137970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC137971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC145068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC079583.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238734 BC088988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P65703 BC137970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P65703 BC137971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P65703 BC145068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P65703 BC079583.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P65703 BC088988.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114948 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09483422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28161778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09285515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09248838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09483437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09481691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09481690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09248839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09483423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09106273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09671391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09248837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28161694 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382855 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382857 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382856 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382861 - -
          Affymetrix HT MG-430 PM Array Plate 1455487_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457836_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441602_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427815_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382852 - -
          Mouse Genome 430 2.0 Array 1455487_at - -
          Mouse Genome 430 2.0 Array 1455487_PM_at - -
          Mouse Genome 430 2.0 Array 1457836_PM_at - -
          Mouse Genome 430 2.0 Array 1457836_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69900_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69900_at - -
          Mouse Expression Set 430 1455487_at - -
          Mouse Expression Set 430 1457836_PM_at - -
          Mouse Expression Set 430 1455487_PM_at - -
          Mouse Expression Set 430 1457836_at - -
          Affymetrix HT Mouse Genome 430A Array 1427815_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427815_at - -
          Affymetrix HT Mouse Genome 430B Array 1457836_at - -
          Affymetrix HT Mouse Genome 430B Array 1455487_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457836_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455487_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103140195 - -
          MouseWG-6 v2 Expression BeadChip 130576 - -
          Illumina mouse-6 v1.0 expression beadchip 70026 - -
          Illumina mouse-6 v1.0 expression beadchip 130576 - -
          Illumina mouse-6 v1.0 expression beadchip 103140195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000130576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673468 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2682173 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2682173 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673468 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219213 - -
          mouse-6 v1.1 (Illumina) scl0069900.1_163-S - -
          mouse-6 v1.1 (Illumina) scl069900.3_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 103140195 - -
          Illumina Sentrix mouse-6.v1 bead array 130576 - -
          Illumina Sentrix mouse-6.v1 bead array 70026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl069900.3_8-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069900.1_163-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70026 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103140195 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 130576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2682173 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216901 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219213 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2682173 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216901 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl069900.3_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline