Search
Search
| Gene Symbol: Gnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008142.4 | NP_032168.1 | 11 - 12 | 1346 | 66 | |
| GenBank mRNA | AB246709.1 | - | 9 - 10 | 1040 | 66 | |
| AK163844.1 | - | 11 - 12 | 1335 | 66 | ||
| AK190349.1 | - | 2 - 3 | 191 | 66 | ||
Set Membership: |
|