Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbxo44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104375.1 | 4 - 5 | 647 | 99 | ||
GenBank mRNA | BC028884.1 | - | 5 - 6 | 791 | 4165174 | 99 |
CX202860.1 | - | 4 - 5 | 423 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |