Search
Search

Ppp5c| Gene Symbol: Ppp5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011155.2 | NP_035285.2 | 5 - 6 | 803 | 134 | |
| GenBank mRNA | AF018262.1 | - | 5 - 6 | 774 | 134 | |
| BC003744.1 | - | 5 - 6 | 759 | 3590322 | 134 | |
Set Membership: |
|