Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264629_m1
          See other 9530026F06RIK GE Assays ›
          Gene Symbol:
          9530026F06Rik
          Gene Name:
          RIKEN cDNA 9530026F06 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 61378432 - 61395904 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264629_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9530026F06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040483.1 3 - 4 513 168
          GenBank mRNA AK020574.1 - 3 - 4 514 168

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000493.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351493 AF017639.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351493 AK019509.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351493 AK038874.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351493 BC021444.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351493 NM_018867.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375614 AB072498.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375614 AK033489.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375614 AK050858.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375614 AK088918.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375614 BC053107.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375614 BC078633.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375614 NM_029943.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410263 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00081682 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100027 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00081682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01076207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00440093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00067074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01076214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00067053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01005171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00651085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20274619 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100004046.mm.1 NR_040483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100004046.mm.1 NR_040483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100004046.mm.1 TC0100000493.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100004045.mm.1 NR_040483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100004045.mm.1 NR_040483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100004045.mm.1 TC0100000493.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 736873 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101850086 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline