Search
Search

Riok3| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024182.4 | NP_077144.2 | 8 - 9 | 1079 | 122 | |
| GenBank mRNA | AK004748.1 | - | 8 - 9 | 1171 | 122 | |
| AK011110.1 | - | 1 - 2 | 163 | 122 | ||
| AK131635.1 | - | 8 - 9 | 1083 | 122 | ||
| AK134327.1 | - | 8 - 9 | 1171 | 122 | ||
| AK147074.1 | - | 8 - 9 | 1080 | 122 | ||
| AK167309.1 | - | 8 - 9 | 1175 | 122 | ||
| AK170882.1 | - | 8 - 9 | 1159 | 122 | ||
| BC002255.1 | - | 5 - 6 | 609 | 122 | ||
| BC033271.1 | - | 8 - 9 | 1117 | 4016197 | 122 | |
Set Membership: |
|