Search
Search

Gucy2g| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081076.2 | NP_001074545.1 | 17 - 18 | 2835 | 70 | |
| GenBank mRNA | AK010727.1 | - | 2 - 3 | 272 | 70 | |
| AK049940.1 | - | 5 - 6 | 854 | 70 | ||
| AY395631.1 | - | 17 - 18 | 2835 | 70 | ||
Set Membership: |
|