Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01265500_m1
          See other L1TD1 GE Assays ›
          Gene Symbol:
          L1td1
          Gene Name:
          LINE-1 type transposase domain containing 1
          Gene Aliases:
          AA546746, AB211064, D76865, ECAT11
          Chromosome Location:
          Chr.4: 98726754 - 98738486 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01265500_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: L1td1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081202.1 NP_001074671.1 3 - 4 1100 90
          GenBank mRNA AB211064.1 - 3 - 4 1106 90
          AK145814.1 - 3 - 4 1093 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          viral or transposable element protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101526 AB211064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101526 NM_001081202.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141942 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159053 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101526 AB211064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101526 NM_001081202.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159053 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05330255 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23140301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03496055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03622644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03622634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03927429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03927426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03622632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03824182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03986295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03824185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03824184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04031263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03332516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03622635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03622630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03696045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03986303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03622643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03622637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03986304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03536195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23140218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03986298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10506113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506112 - -
          Affymetrix HT MG-430 PM Array Plate 1457314_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10506110 - -
          Mouse Genome 430 2.0 Array 1457314_PM_at - -
          Mouse Genome 430 2.0 Array 1457314_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381591_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381591_at - -
          Affymetrix Mouse Clariom S Array PSR0400006659.mm.1 NM_001081202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006659.mm.1 NM_001081202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006659.mm.1 TC0400000987.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006662.mm.1 NM_001081202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006662.mm.1 NM_001081202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006662.mm.1 TC0400000987.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006660.mm.1 NM_001081202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006660.mm.1 NM_001081202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006660.mm.1 TC0400000987.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006664.mm.1 NM_001081202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006664.mm.1 NM_001081202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006664.mm.1 TC0400000987.mm.1 MTA TC
          Mouse Expression Set 430 1457314_at - -
          Mouse Expression Set 430 1457314_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457314_at - -
          Affymetrix HT Mouse Genome 430B Array 1457314_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 675545 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260018 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2961042 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2961042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline