Search
Search
| Gene Symbol: Bcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029933.4 | NP_084209.3 | 9 - 10 | 3780 | 71 | |
| GenBank mRNA | AK084676.1 | - | 2 - 3 | 1707 | 71 | |
| AK147659.1 | - | 9 - 10 | 3780 | 71 | ||
| AY296061.1 | - | 6 - 7 | 3296 | 71 | ||
| BC019641.1 | - | 1 - 2 | 227 | 71 | ||
| BC137850.1 | - | 6 - 7 | 3268 | 8860869 | 71 | |
Set Membership: |
|