Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01265768_g1
          See other ITGA10 GE Assays ›
          Gene Symbol:
          Itga10
          Gene Name:
          integrin, alpha 10
          Gene Aliases:
          A630048L14
          Chromosome Location:
          Chr.3: 96645584 - 96664519 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01265768_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itga10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK037222.1 - 9 - 10 801 94
          BC115770.1 - 29 - 30 3469 40085898 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003218.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          integrin

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097913 NM_001081053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111533 NM_001081053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111533 AK037222.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46209 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132597 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097913 NM_001081053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111533 NM_001081053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111533 AK037222.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04998256 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132597 - -
          Whole Mouse Genome G4122A A_51_P231697 AK037222.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02361583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02814037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02814041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02555723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02426273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02361580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02888018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02426295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02306229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03147813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02306235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02888017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02927611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02426261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02695739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350404 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494499 - -
          Affymetrix HT MG-430 PM Array Plate 1440235_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10494467 - -
          Mouse Genome 430 2.0 Array 1440235_at - -
          Mouse Genome 430 2.0 Array 1440235_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 213119_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 213119_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300007633.mm.1 TC0300003218.mm.1 MTA TC
          Mouse Expression Set 430 1440235_at - -
          Mouse Expression Set 430 1440235_PM_at - -
          Murine Genome U74v2 Set 135131_at - -
          Murine Genome U74v2 Set 135131_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440235_at - -
          Affymetrix HT Mouse Genome 430B Array 1440235_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692244 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692244 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline