Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267260_s1
          See other MC4R GE Assays ›
          Gene Symbol:
          Mc4r
          Gene Name:
          melanocortin 4 receptor
          Gene Aliases:
          Mc4-r
          Chromosome Location:
          Chr.18: 66857705 - 66860487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267260_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mc4r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016977.4 NP_058673.2 1 1365 100
          GenBank mRNA AK136793.1 - 1 1351 100
          BC116957.1 - 1 983 40126276 100
          BC116959.1 - 1 983 40126278 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mc4r

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          diet induced thermogenesis
          energy reserve metabolic process
          signal transduction
          G-protein coupled receptor signaling pathway
          feeding behavior
          regulation of metabolic process
          insulin secretion
          positive regulation of cAMP biosynthetic process
          response to insulin
          positive regulation of bone resorption
          regulation of feeding behavior
          negative regulation of feeding behavior
          regulation of grooming behavior
          signal transducer activity
          G-protein coupled receptor activity
          melanocortin receptor activity
          melanocyte-stimulating hormone receptor activity
          protein binding
          peptide hormone binding
          ubiquitin protein ligase binding
          neuropeptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183884 AK136793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183884 BC116957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183884 NM_016977.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185714 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122164 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160650 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147350 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183884 AK136793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183884 BC116957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183884 NM_016977.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268709 AK136793.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268709 BC116957.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268709 BC116959.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268709 NM_016977.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788800 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122164 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147350 - -
          Whole Mouse Genome G4122A A_51_P268709 AK136793.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268709 BC116957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268709 BC116959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268709 NM_016977.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117217 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122164 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147350 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160650 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102913 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13963830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13963845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13976333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14102955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14000913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14000912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788780 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10459517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459516 - -
          Affymetrix Mouse Gene 1.1 ST Array 10459512 - -
          Affymetrix Mouse Clariom S Array PSR1800010950.mm.1 NM_016977.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010950.mm.1 TC1800001485.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 366456 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 430333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430333 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2611237 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2611237 - -
          mouse-6 v1.1 (Illumina) scl14872.1.1_153-S - -
          Illumina Sentrix mouse-6.v1 bead array 430333 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl14872.1.1_153-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430333 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2611237 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2611237 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline