Search
Search

Zyg11a| Gene Symbol: Zyg11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167936.1 | NP_001161408.1 | 11 - 12 | 1943 | 68 | |
| GenBank mRNA | BC022150.1 | - | 9 - 10 | 1491 | 68 | |
Set Membership: |
|