Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01267930_m1
See other SLCO1B2 GE Assays ›
Gene Symbol:
Slco1b2
Gene Name:
solute carrier organic anion transporter family, member 1b2
Gene Aliases:
7330442B20Rik, OATP-C, OATP2, Oatp1b2, Oatp4, Slc21a10, Slc21a6, lst-1, mlst-1
Chromosome Location:
Chr.6: 141629452 - 141686646 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01267930_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Slco1b2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_020495.1
NP_065241.1
2 - 3
124
88
GenBank mRNA
AB031959.1
-
2 - 3
124
88
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the AB031959.1, NM_020495.1 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Transporter
Mus musculus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Slco1b2
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
drug transmembrane transport
oligopeptide transport
organic anion transport
bile acid and bile salt transport
drug transport
anion transmembrane transport
organic acid transmembrane transport
transporter activity
organic anion transmembrane transporter activity
bile acid transmembrane transporter activity
oligopeptide transporter activity
drug transmembrane transporter activity
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR0600013323.mm.1
TC0600001672.mm.1
MTA TC
Affymetrix Mouse Clariom S Array
PSR0600013323.mm.1
NM_020495.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0600013323.mm.1
NM_020495.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0600013324.mm.1
TC0600001672.mm.1
MTA TC
Affymetrix Mouse Clariom S Array
PSR0600013324.mm.1
NM_020495.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0600013324.mm.1
NM_020495.1
Refseq
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline