Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268318_s1
          See other C5AR1 GE Assays ›
          Gene Symbol:
          C5ar1
          Gene Name:
          complement component 5a receptor 1
          Gene Aliases:
          C5aR, C5r1, Cd88, D7Msu1
          Chromosome Location:
          Chr.7: 16246743 - 16259540 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268318_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C5ar1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001173550.1 NP_001167021.1 2 1199 72
          NM_007577.4 NP_031603.2 3 1138 72
          GenBank mRNA AK150407.1 - 1 858 72
          AK158027.1 - 2 1058 72
          AY220494.1 - 1 1008 72
          BC125641.1 - 2 1199 40130156 72
          BC125643.1 - 2 1199 40130158 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C5ar1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          complement receptor mediated signaling pathway
          apoptotic process
          chemotaxis
          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          positive regulation of vascular endothelial growth factor production
          positive regulation of macrophage chemotaxis
          cell proliferation in hindbrain
          neutrophil chemotaxis
          response to peptidoglycan
          complement component C5a signaling pathway
          mRNA transcription from RNA polymerase II promoter
          negative regulation of neuron apoptotic process
          positive regulation of angiogenesis
          positive regulation of epithelial cell proliferation
          defense response to Gram-positive bacterium
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of neutrophil chemotaxis
          complement component C5a binding
          signal transducer activity
          complement component C5a receptor activity
          G-protein coupled receptor activity
          anaphylatoxin receptor activity
          C5a anaphylatoxin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017681 NM_001173550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017681 NM_007577.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16088 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178612 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017681 NM_001173550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017681 NM_007577.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279542 AK158027.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279542 AY220494.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279542 BC125641.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279542 BC125643.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279542 NM_001173550.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980480 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178612 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06129078 - -
          Whole Mouse Genome G4122A A_51_P279542 AK158027.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279542 AY220494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279542 BC125641.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279542 BC125643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279542 NM_001173550.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118289 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980527 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560244 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560246 - -
          Affymetrix HT MG-430 PM Array Plate 1439902_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422190_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560242 - -
          Mouse Genome 430 2.0 Array 1422190_PM_at - -
          Mouse Genome 430 2.0 Array 1439902_PM_at - -
          Mouse Genome 430 2.0 Array 1439902_at - -
          Mouse Genome 430 2.0 Array 1422190_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12273_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12273_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700021315.mm.1 TC0700002422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021315.mm.1 NM_001173550.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021315.mm.1 NM_001173550.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021315.mm.1 NM_007577.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021315.mm.1 NM_007577.4 Refseq
          Mouse Expression Set 430 1422190_at - -
          Mouse Expression Set 430 1422190_PM_at - -
          Mouse Expression Set 430 1439902_PM_at - -
          Mouse Expression Set 430 1439902_at - -
          Murine Genome U74v2 Set 101728_PM_at - -
          Murine Genome U74v2 Set 101728_at - -
          Mouse Genome 430A 2.0 Array 1422190_PM_at - -
          Mouse Genome 430A 2.0 Array 1422190_at - -
          Affymetrix HT Mouse Genome 430A Array 1422190_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422190_at - -
          Affymetrix HT Mouse Genome 430B Array 1439902_at - -
          Affymetrix HT Mouse Genome 430B Array 1439902_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 338868 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4540402 - -
          Illumina mouse-6 v1.0 expression beadchip 4540402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2595052 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2595052 - -
          mouse-6 v1.1 (Illumina) scl31725.3.3_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 4540402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31725.3.3_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2595052 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2595052 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline