Search
Search

Sirpa| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291019.1 | NP_001277948.1 | 3 - 4 | 1197 | 66 | |
| NM_001291020.1 | NP_001277949.1 | 4 - 5 | 999 | 66 | ||
| NM_007547.4 | NP_031573.2 | 3 - 4 | 1197 | 66 | ||
| GenBank mRNA | AB018194.1 | - | 3 - 4 | 763 | 66 | |
| AF072543.1 | - | 3 - 4 | 753 | 66 | ||
| AF332079.1 | - | 3 - 4 | 763 | 66 | ||
| AF332080.1 | - | 3 - 4 | 763 | 66 | ||
| AK193777.1 | - | 1 - 2 | 219 | 66 | ||
| AK197651.1 | - | 1 - 2 | 219 | 66 | ||
| BC062197.1 | - | 3 - 4 | 788 | 5368250 | 66 | |
| D87967.1 | - | 3 - 4 | 1162 | 66 | ||
| D87968.1 | - | 3 - 4 | 1162 | 66 | ||
| U89694.1 | - | 3 - 4 | 774 | 66 | ||
| Y10349.1 | - | 3 - 4 | 808 | 66 | ||
Set Membership: |
|