Search
Search

Tnnt3| Gene Symbol: Tnnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163664.1 | NP_001157136.1 | 8 - 9 | 189 | 119 | |
| NM_001163665.1 | NP_001157137.1 | 8 - 9 | 189 | 119 | ||
| NM_001163667.1 | NP_001157139.1 | 7 - 8 | 177 | 107 | ||
| GenBank mRNA | AI574395.1 | - | 8 - 9 | 183 | 119 | |
| BC003747.1 | - | 8 - 9 | 185 | 3594256 | 119 | |
| BY766323.1 | - | 7 - 8 | 167 | 107 | ||
| L49466.1 | - | 7 - 8 | 177 | 107 | ||
| L49467.1 | - | 8 - 9 | 189 | 119 | ||
| L49471.1 | - | 6 - 7 | 159 | 107 | ||
Set Membership: |
|