Search
Search

Fchsd2| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146010.1 | NP_001139482.1 | 13 - 14 | 1552 | 108 | |
| NM_199012.2 | NP_950177.2 | 14 - 15 | 1624 | 108 | ||
| GenBank mRNA | AK122368.1 | - | 14 - 15 | 1516 | 108 | |
| AK140322.1 | - | 13 - 14 | 1531 | 108 | ||
| AK141949.1 | - | 13 - 14 | 1552 | 108 | ||
| BC034086.1 | - | 1 - 2 | 99 | 108 | ||
| BC060647.1 | - | 14 - 15 | 1586 | 6854041 | 108 | |
Set Membership: |
|