Search
Search

Aars2| Gene Symbol: Aars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198608.2 | NP_941010.2 | 19 - 20 | 2596 | 62 | |
| GenBank mRNA | AK146958.1 | - | 19 - 20 | 2597 | 62 | |
| AK167771.1 | - | 19 - 20 | 2591 | 62 | ||
| AK173139.1 | - | 15 - 16 | 1870 | 62 | ||
| BC059889.1 | - | 10 - 11 | 1625 | 62 | ||
| BC079844.1 | - | 19 - 20 | 2568 | 62 | ||
| BC110699.1 | - | 10 - 11 | 1625 | 62 | ||
| BC113172.1 | - | 19 - 20 | 2592 | 40060953 | 62 | |
| BC113779.1 | - | 19 - 20 | 2592 | 40060952 | 62 | |
Set Membership: |
|