Search
Search

Pcsk2| Gene Symbol: Pcsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008792.4 | NP_032818.1 | 4 - 5 | 859 | 63 | |
| GenBank mRNA | AF483513.1 | - | 4 - 5 | 501 | 63 | |
| AK133620.1 | - | 4 - 5 | 860 | 63 | ||
| AK158493.1 | - | 4 - 5 | 807 | 63 | ||
| AK158598.1 | - | 4 - 5 | 921 | 63 | ||
| AK161585.1 | - | 4 - 5 | 800 | 63 | ||
| BC052013.1 | - | 4 - 5 | 776 | 5707923 | 63 | |
| BC057348.1 | - | 4 - 5 | 814 | 6827695 | 63 | |
| M55669.1 | - | 4 - 5 | 585 | 63 | ||
Set Membership: |
|