Search
Search
| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172671.2 | NP_766259.2 | 5 - 6 | 1089 | 104 | |
| GenBank mRNA | AK044357.1 | - | 4 - 5 | 397 | 104 | |
| AK199068.1 | - | 1 - 2 | 102 | 104 | ||
| CF164628.1 | - | 1 - 2 | 111 | 104 | ||
Set Membership: |
|