Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01270578_g1
          See other SLC29A1 GE Assays ›
          Gene Symbol:
          Slc29a1
          Gene Name:
          solute carrier family 29 (nucleoside transporters), member 1
          Gene Aliases:
          1200014D21Rik, AA407560, ENT1
          Chromosome Location:
          Chr.17: 45585200 - 45599608 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01270578_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc29a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199113.1 NP_001186042.1 11 - 12 1211 69
          NM_001199114.1 NP_001186043.1 10 - 11 1050 69
          NM_001199115.1 NP_001186044.1 10 - 11 1150 69
          NM_001199116.1 NP_001186045.1 10 - 11 1110 69
          NM_022880.3 NP_075018.1 9 - 10 1151 69
          GenBank mRNA AA407561.1 - 4 - 5 317 69
          AF131212.1 - 9 - 10 1152 69
          AF305501.1 - 10 - 11 1121 69
          AK004756.1 - 10 - 11 1096 69
          AK050089.1 - 10 - 11 1135 69
          AK146265.1 - 10 - 11 1051 69
          AK149483.1 - 10 - 11 1066 69
          AK170463.1 - 10 - 11 1082 69
          AK198320.1 - 1 - 2 97 69
          AK212529.1 - 1 - 2 97 69
          BC004828.1 - 11 - 12 1174 2631728 69
          BC006812.1 - 9 - 10 1082 3155188 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002152.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc29a1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          nucleoside transport
          uridine transport
          sleep
          excitatory postsynaptic potential
          nucleoside transmembrane transport
          nucleoside transmembrane transporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_001199113.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_001199113.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_001199114.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_001199114.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_001199115.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_001199115.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_001199116.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_001199116.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_022880.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 NM_022880.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020116.mm.1 TC1700002152.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline