Search
Search

Fnbp4| Gene Symbol: Fnbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018828.2 | NP_061298.1 | 4 - 5 | 872 | 71 | |
| GenBank mRNA | AK012167.1 | - | 4 - 5 | 685 | 71 | |
| AK014018.1 | - | 4 - 5 | 689 | 71 | ||
| AK084018.1 | - | 4 - 5 | 664 | 71 | ||
| AK129263.1 | - | 4 - 5 | 769 | 71 | ||
| AK164543.1 | - | 4 - 5 | 643 | 71 | ||
| U40750.2 | - | 4 - 5 | 872 | 71 | ||
Set Membership: |
|