Search
Search

Stk39| Gene Symbol: Stk39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016866.2 | NP_058562.1 | 12 - 13 | 1383 | 101 | |
| GenBank mRNA | AF099988.1 | - | 12 - 13 | 1359 | 101 | |
| AK017292.1 | - | 12 - 13 | 1343 | 101 | ||
| AK132016.1 | - | 5 - 6 | 716 | 101 | ||
| AK148456.1 | - | 12 - 13 | 1420 | 101 | ||
| AK163194.1 | - | 2 - 3 | 98 | 101 | ||
| BC051964.1 | - | 12 - 13 | 1670 | 5698326 | 101 | |
| BC064443.1 | - | 12 - 13 | 1383 | 6843981 | 101 | |
| JN368425.1 | - | 8 - 9 | 964 | 101 | ||
Set Membership: |
|