Search
Search

Stk39| Gene Symbol: Stk39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016866.2 | NP_058562.1 | 16 - 17 | 1634 | 111 | |
| GenBank mRNA | AF099988.1 | - | 16 - 17 | 1610 | 111 | |
| AK132016.1 | - | 9 - 10 | 967 | 111 | ||
| AK148456.1 | - | 16 - 17 | 1671 | 111 | ||
| AK163194.1 | - | 6 - 7 | 349 | 111 | ||
| BC051964.1 | - | 16 - 17 | 1921 | 5698326 | 111 | |
| BC064443.1 | - | 16 - 17 | 1634 | 6843981 | 111 | |
| JN368425.1 | - | 12 - 13 | 1215 | 111 | ||
Set Membership: |
|