| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200030567.mm.1 |
NM_001159636.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030567.mm.1 |
NM_001159636.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030567.mm.1 |
NM_028118.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030567.mm.1 |
NM_028118.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030567.mm.1 |
TC0200003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200030560.mm.1 |
NM_001159636.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030560.mm.1 |
NM_001159636.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030560.mm.1 |
NM_028118.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030560.mm.1 |
NM_028118.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030560.mm.1 |
TC0200003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200030562.mm.1 |
NM_001159636.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030562.mm.1 |
NM_001159636.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030562.mm.1 |
NM_028118.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030562.mm.1 |
NM_028118.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030562.mm.1 |
TC0200003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200030565.mm.1 |
NM_001159636.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030565.mm.1 |
NM_001159636.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030565.mm.1 |
NM_028118.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030565.mm.1 |
NM_028118.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030565.mm.1 |
TC0200003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200030566.mm.1 |
NM_001159636.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030566.mm.1 |
NM_001159636.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030566.mm.1 |
NM_028118.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030566.mm.1 |
NM_028118.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200030566.mm.1 |
TC0200003590.mm.1 |
MTA TC |