Search
Search
| Gene Symbol: Klc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029091.2 | NP_083367.1 | 4 - 5 | 637 | 110 | |
| GenBank mRNA | AK004774.1 | - | 4 - 5 | 638 | 110 | |
| BC005746.1 | - | 4 - 5 | 659 | 3599898 | 110 | |
Set Membership: |
|