Search
Search

Dctn5| Gene Symbol: Dctn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021608.3 | NP_067621.3 | 5 - 6 | 547 | 119 | |
| GenBank mRNA | AF190795.1 | - | 5 - 6 | 518 | 119 | |
| AK010238.1 | - | 5 - 6 | 527 | 119 | ||
| AK051957.1 | - | 5 - 6 | 486 | 119 | ||
| AK076100.1 | - | 5 - 6 | 520 | 119 | ||
| AK076738.1 | - | 5 - 6 | 547 | 119 | ||
| AK145958.1 | - | 4 - 5 | 1498 | 119 | ||
| AK146123.1 | - | 5 - 6 | 513 | 119 | ||
| AK151193.1 | - | 5 - 6 | 519 | 119 | ||
| AK151876.1 | - | 5 - 6 | 519 | 119 | ||
| AK168983.1 | - | 5 - 6 | 527 | 119 | ||
| BC005671.1 | - | 4 - 5 | 1521 | 3153175 | 119 | |
| BC010834.1 | - | 5 - 6 | 516 | 4235312 | 119 | |
| BC092273.1 | - | 4 - 5 | 1500 | 119 | ||
Set Membership: |
|