Search
Search

Spns1| Gene Symbol: Spns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023712.3 | NP_076201.2 | 5 - 6 | 1520 | 116 | |
| NR_045537.1 | 6 - 7 | 1499 | 116 | |||
| GenBank mRNA | AF212372.1 | - | 5 - 6 | 1011 | 116 | |
| AK008721.1 | - | 6 - 7 | 996 | 116 | ||
| AK033907.1 | - | 3 - 4 | 2075 | 116 | ||
| AK154353.1 | - | 5 - 6 | 1010 | 116 | ||
| AK166931.1 | - | 5 - 6 | 1012 | 116 | ||
| BC002297.1 | - | 5 - 6 | 539 | 116 | ||
| BC026854.1 | - | 5 - 6 | 981 | 4242049 | 116 | |
| BC085491.1 | - | 5 - 6 | 964 | 6829415 | 116 | |
Set Membership: |
|